Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCGCGTTTACTGAAAAATCATCC[A/G]TTGCATTGGCTCTAAAGGTATCCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605644 MIM: 605504 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KLK8 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KLK8 - kallikrein related peptidase 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281431.1 | 1406 | Intron | NP_001268360.1 | |||
NM_007196.3 | 1406 | Intron | NP_009127.1 | |||
NM_144505.2 | 1406 | Intron | NP_653088.1 | |||
NM_144506.2 | 1406 | Intron | NP_653089.1 | |||
NM_144507.2 | 1406 | Intron | NP_653090.1 |
KLK9 - kallikrein related peptidase 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012315.1 | 1406 | UTR 3 | NP_036447.1 |
Set Membership: |
HapMap |