Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609290 MIM: 610346 | ||||||||||||||||||||
Literature Links: |
AK3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AK3 - adenylate kinase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199852.1 | 1931 | UTR 3 | NP_001186781.1 | |||
NM_001199853.1 | 1931 | UTR 3 | NP_001186782.1 | |||
NM_001199855.1 | 1931 | UTR 3 | NP_001186784.1 | |||
NM_001199856.1 | 1931 | UTR 3 | NP_001186785.1 | |||
NM_016282.3 | 1931 | UTR 3 | NP_057366.2 |
CDC37L1 - cell division cycle 37 like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |