Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAGAGAGTTCAAGGAGGCTCTGC[A/G]TCTCCATGTCTCCTGCCCTGCTCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
UBE2F PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
UBE2F - ubiquitin conjugating enzyme E2 F (putative) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278305.1 | Intron | NP_001265234.1 | ||||
NM_001278306.1 | Intron | NP_001265235.1 | ||||
NM_001278307.1 | Intron | NP_001265236.1 | ||||
NM_001278308.1 | Intron | NP_001265237.1 | ||||
NM_080678.2 | Intron | NP_542409.1 |
UBE2F-SCLY - UBE2F-SCLY readthrough (NMD candidate) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |