Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGAGACACAGGGAAGAACTGTGCG[A/C]GTTAGAGGTGGGGAGGAGGAAGGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105377805 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
LOC105377805 - basic salivary proline-rich protein 4-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM255B - transmembrane protein 255B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_182614.2 | Intron | NP_872420.1 | ||||
XM_005272413.3 | Intron | XP_005272470.1 | ||||
XM_005272415.2 | Intron | XP_005272472.1 | ||||
XM_006719986.2 | Intron | XP_006720049.1 | ||||
XM_017020558.1 | Intron | XP_016876047.1 | ||||
XM_017020559.1 | Intron | XP_016876048.1 | ||||
XM_017020560.1 | Intron | XP_016876049.1 |