Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGAGGGATGCTTAATTTAAACTCC[A/C]AAAAAGCTCATTTTTTTGCAAAGCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605501 | ||||||||||||||||||||
Literature Links: |
MPHOSPH9 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MPHOSPH9 - M-phase phosphoprotein 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022782.3 | 4756 | Intron | NP_073619.3 | |||
XM_011537740.2 | 4756 | UTR 3 | XP_011536042.1 | |||
XM_011537741.2 | 4756 | Intron | XP_011536043.1 | |||
XM_017018673.1 | 4756 | Intron | XP_016874162.1 | |||
XM_017018674.1 | 4756 | Intron | XP_016874163.1 | |||
XM_017018675.1 | 4756 | Intron | XP_016874164.1 | |||
XM_017018676.1 | 4756 | Intron | XP_016874165.1 | |||
XM_017018677.1 | 4756 | Intron | XP_016874166.1 | |||
XM_017018678.1 | 4756 | Intron | XP_016874167.1 | |||
XM_017018679.1 | 4756 | Intron | XP_016874168.1 | |||
XM_017018680.1 | 4756 | Intron | XP_016874169.1 | |||
XM_017018681.1 | 4756 | Intron | XP_016874170.1 |