Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609642 MIM: 615450 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TARP PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
TARP - TCR gamma alternate reading frame protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003799.1 | Intron | NP_001003799.1 | ||||
NM_001003806.1 | Intron | NP_001003806.1 | ||||
XM_011515407.2 | Intron | XP_011513709.1 | ||||
XM_017012226.1 | Intron | XP_016867715.1 | ||||
XM_017012227.1 | Intron | XP_016867716.1 | ||||
XM_017012228.1 | Intron | XP_016867717.1 | ||||
XM_017012229.1 | Intron | XP_016867718.1 | ||||
XM_017012230.1 | Intron | XP_016867719.1 |
TRGC2 - T cell receptor gamma constant 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |