Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCTTTTGCTGACCATCTGCCATG[C/T]AGTAAGGGCAGAGGGCTGTCTTATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
15 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600650 MIM: 602603 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1orf123 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C1orf123 - chromosome 1 open reading frame 123 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304759.1 | Intron | NP_001291688.1 | ||||
NM_001304760.1 | Intron | NP_001291689.1 | ||||
NM_017887.2 | Intron | NP_060357.1 |
CPT2 - carnitine palmitoyltransferase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MAGOH - mago homolog, exon junction complex core component | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |