Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGGAATGGACTGAGTTGGGGTGG[A/G]GGCCGGGTTTCCTTTCCCTGGCTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600328 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105371763 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC105371763 - uncharacterized LOC105371763 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR4734 - microRNA 4734 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MLLT6 - MLLT6, PHD finger domain containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005937.3 | Intron | NP_005928.2 | ||||
XM_006721911.2 | Intron | XP_006721974.1 | ||||
XM_017024648.1 | Intron | XP_016880137.1 | ||||
XM_017024649.1 | Intron | XP_016880138.1 | ||||
XM_017024650.1 | Intron | XP_016880139.1 |
Set Membership: |
HapMap |