Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611947 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NLRX1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NLRX1 - NLR family member X1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282143.1 | 668 | Missense Mutation | CCC,TCC | P,S 63 | NP_001269072.1 | |
NM_001282144.1 | 668 | Missense Mutation | CCC,TCC | P,S 63 | NP_001269073.1 | |
NM_001282358.1 | 668 | Missense Mutation | CCC,TCC | P,S 63 | NP_001269287.1 | |
NM_024618.3 | 668 | Missense Mutation | CCC,TCC | P,S 63 | NP_078894.2 | |
XM_005271669.1 | 668 | Missense Mutation | CCC,TCC | P,S 63 | XP_005271726.1 | |
XM_005271670.1 | 668 | UTR 5 | XP_005271727.1 | |||
XM_005271671.1 | 668 | UTR 5 | XP_005271728.1 | |||
XM_005271672.1 | 668 | UTR 5 | XP_005271729.1 | |||
XM_006718904.1 | 668 | Missense Mutation | CCC,TCC | P,S 63 | XP_006718967.1 | |
XM_011542980.1 | 668 | Missense Mutation | CCC,TCC | P,S 63 | XP_011541282.1 | |
XM_017018294.1 | 668 | Missense Mutation | CCC,TCC | P,S 63 | XP_016873783.1 |
Set Membership: |
HapMap |