Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGCCCTGCTACACTCACTGCTTA[A/G]CAGACGTCAGACTGGTGGTCGTCTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610851 | ||||||||||||||||||||
Literature Links: |
AP1AR PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AP1AR - adaptor related protein complex 1 associated regulatory protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128426.2 | 49 | Intron | NP_001121898.1 | |||
NM_018569.5 | 49 | Intron | NP_061039.3 | |||
XM_017008378.1 | 49 | Intron | XP_016863867.1 | |||
XM_017008379.1 | 49 | Intron | XP_016863868.1 | |||
XM_017008380.1 | 49 | UTR 5 | XP_016863869.1 |
LOC105377371 - uncharacterized LOC105377371 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |