Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
53 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611471 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACP6 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ACP6 - acid phosphatase 6, lysophosphatidic | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001323625.1 | Intron | NP_001310554.1 | ||||
NM_016361.4 | Intron | NP_057445.4 | ||||
XM_011509601.2 | Intron | XP_011507903.1 | ||||
XM_017001422.1 | Intron | XP_016856911.1 |
Set Membership: |
HapMap |