Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
50 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614772 MIM: 605052 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
COA6 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
COA6 - cytochrome c oxidase assembly factor 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TARBP1 - TAR (HIV-1) RNA binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005646.3 | 2900 | Silent Mutation | GAC,GAT | D,D 1429 | NP_005637.3 | |
XM_005273234.3 | 2900 | Silent Mutation | GAC,GAT | D,D 1428 | XP_005273291.1 | |
XM_011544263.2 | 2900 | Silent Mutation | GAC,GAT | D,D 1405 | XP_011542565.1 | |
XM_017002194.1 | 2900 | Intron | XP_016857683.1 | |||
XM_017002195.1 | 2900 | Silent Mutation | GAC,GAT | D,D 973 | XP_016857684.1 | |
XM_017002196.1 | 2900 | Silent Mutation | GAC,GAT | D,D 686 | XP_016857685.1 | |
XM_017002197.1 | 2900 | Silent Mutation | GAC,GAT | D,D 632 | XP_016857686.1 |
Set Membership: |
HapMap JSNP |