Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
10 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
SPATA21 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SPATA21 - spermatogenesis associated 21 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198546.1 | 1842 | Intron | NP_940948.1 | |||
XM_011541407.2 | 1842 | Intron | XP_011539709.1 | |||
XM_011541417.2 | 1842 | Intron | XP_011539719.1 | |||
XM_017001216.1 | 1842 | Intron | XP_016856705.1 | |||
XM_017001217.1 | 1842 | Intron | XP_016856706.1 | |||
XM_017001218.1 | 1842 | Intron | XP_016856707.1 | |||
XM_017001219.1 | 1842 | Intron | XP_016856708.1 | |||
XM_017001220.1 | 1842 | Intron | XP_016856709.1 | |||
XM_017001221.1 | 1842 | Intron | XP_016856710.1 | |||
XM_017001222.1 | 1842 | Intron | XP_016856711.1 | |||
XM_017001223.1 | 1842 | Intron | XP_016856712.1 | |||
XM_017001224.1 | 1842 | Intron | XP_016856713.1 | |||
XM_017001225.1 | 1842 | Intron | XP_016856714.1 |
SZRD1 - SUZ RNA binding domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001114600.2 | 1842 | UTR 3 | NP_001108072.1 | |||
NM_001271869.1 | 1842 | UTR 3 | NP_001258798.1 |