Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610668 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
INSC PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
INSC - inscuteable homolog (Drosophila) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031853.4 | Intron | NP_001027024.3 | ||||
NM_001042536.2 | Intron | NP_001036001.1 | ||||
NM_001278313.1 | Intron | NP_001265242.1 | ||||
NM_001278314.1 | Intron | NP_001265243.1 | ||||
NM_001278315.1 | Intron | NP_001265244.1 | ||||
NM_001278316.1 | Intron | NP_001265245.1 | ||||
XM_006718227.3 | Intron | XP_006718290.1 | ||||
XM_011520085.1 | Intron | XP_011518387.1 | ||||
XM_011520086.1 | Intron | XP_011518388.1 | ||||
XM_017017697.1 | Intron | XP_016873186.1 | ||||
XM_017017698.1 | Intron | XP_016873187.1 |