Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607946 MIM: 608171 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADIPOR2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ADIPOR2 - adiponectin receptor 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CACNA2D4 - calcium voltage-gated channel auxiliary subunit alpha2delta 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_172364.4 | 5032 | UTR 3 | NP_758952.4 | |||
XM_011521041.2 | 5032 | Intron | XP_011519343.1 |
Set Membership: |
HapMap |