Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607719 MIM: 191043 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SYT8 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SYT8 - synaptotagmin 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TNNI2 - troponin I2, fast skeletal type | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145829.1 | Intron | NP_001139301.1 | ||||
NM_001145841.1 | Intron | NP_001139313.1 | ||||
NM_003282.3 | Intron | NP_003273.1 |
Set Membership: |
HapMap |