Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605580 MIM: 600556 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IL23A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IL23A - interleukin 23 subunit alpha | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PAN2 - PAN2 poly(A) specific ribonuclease subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STAT2 - signal transducer and activator of transcription 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005419.3 | 2611 | UTR 3 | NP_005410.1 | |||
NM_198332.1 | 2611 | UTR 3 | NP_938146.1 | |||
XM_005269111.3 | 2611 | Intron | XP_005269168.2 | |||
XM_011538697.2 | 2611 | UTR 3 | XP_011536999.1 | |||
XM_011538698.2 | 2611 | UTR 3 | XP_011537000.1 | |||
XM_011538699.2 | 2611 | Intron | XP_011537001.1 | |||
XM_011538700.2 | 2611 | UTR 3 | XP_011537002.1 | |||
XM_017019904.1 | 2611 | UTR 3 | XP_016875393.1 |