Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611916 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
COL6A5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
COL6A5 - collagen type VI alpha 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278298.1 | 259 | UTR 5 | NP_001265227.1 | |||
NM_153264.6 | 259 | UTR 5 | NP_694996.5 | |||
XM_011512621.2 | 259 | UTR 5 | XP_011510923.1 | |||
XM_011512622.2 | 259 | UTR 5 | XP_011510924.1 | |||
XM_011512623.2 | 259 | UTR 5 | XP_011510925.1 |
Set Membership: |
HapMap |