Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 617029 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SEMA4B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
SEMA4B - semaphorin 4B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001324029.1 | Intron | NP_001310958.1 | ||||
NM_001324030.1 | Intron | NP_001310959.1 | ||||
NM_001324031.1 | Intron | NP_001310960.1 | ||||
NM_001324032.1 | Intron | NP_001310961.1 | ||||
NM_001324033.1 | Intron | NP_001310962.1 | ||||
NM_001324034.1 | Intron | NP_001310963.1 | ||||
NM_020210.3 | Intron | NP_064595.2 | ||||
NM_198925.2 | Intron | NP_945119.1 |
Set Membership: |
HapMap Validated |