Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612136 MIM: 612137 | ||||||||||||||||||||
Literature Links: |
ECHDC1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ECHDC1 - ethylmalonyl-CoA decarboxylase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002030.1 | 2017 | UTR 3 | NP_001002030.1 | |||
NM_001105544.1 | 2017 | UTR 3 | NP_001099014.1 | |||
NM_001105545.1 | 2017 | UTR 3 | NP_001099015.1 | |||
NM_001139510.1 | 2017 | UTR 3 | NP_001132982.1 | |||
NM_018479.3 | 2017 | UTR 3 | NP_060949.2 | |||
XM_005267047.2 | 2017 | UTR 3 | XP_005267104.1 | |||
XM_005267048.1 | 2017 | UTR 3 | XP_005267105.1 | |||
XM_005267049.2 | 2017 | UTR 3 | XP_005267106.1 | |||
XM_005267050.2 | 2017 | UTR 3 | XP_005267107.1 | |||
XM_011535942.2 | 2017 | UTR 3 | XP_011534244.1 | |||
XM_011535943.2 | 2017 | Intron | XP_011534245.1 | |||
XM_017011056.1 | 2017 | UTR 3 | XP_016866545.1 |