Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612675 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FSD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FSD2 - fibronectin type III and SPRY domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007122.3 | 3139 | UTR 3 | NP_001007123.1 | |||
NM_001281805.1 | 3139 | UTR 3 | NP_001268734.1 | |||
NM_001281806.1 | 3139 | UTR 3 | NP_001268735.1 | |||
XM_005272425.4 | 3139 | UTR 3 | XP_005272482.1 | |||
XM_011521235.2 | 3139 | UTR 3 | XP_011519537.1 | |||
XM_017021924.1 | 3139 | Intron | XP_016877413.1 |
SCARNA15 - small Cajal body-specific RNA 15 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNHG21 - small nucleolar RNA host gene 21 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |