Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602121 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DIAPH1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DIAPH1 - diaphanous related formin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001079812.2 | 3693 | Silent Mutation | GGC,GGT | G,G 1184 | NP_001073280.1 | |
NM_001314007.1 | 3693 | Silent Mutation | GGC,GGT | G,G 1193 | NP_001300936.1 | |
NM_005219.4 | 3693 | Silent Mutation | GGC,GGT | G,G 1193 | NP_005210.3 | |
XM_011537572.2 | 3693 | Silent Mutation | GGC,GGT | G,G 1181 | XP_011535874.1 | |
XM_011537573.2 | 3693 | Silent Mutation | GGC,GGT | G,G 1171 | XP_011535875.1 |