Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613009 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SEPSECS PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SEPSECS - Sep (O-phosphoserine) tRNA:Sec (selenocysteine) tRNA synthase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016955.3 | 1543 | Missense Mutation | AAC,AAG | N,K 452 | NP_058651.3 | |
XM_011513846.2 | 1543 | Missense Mutation | AAC,AAG | N,K 451 | XP_011512148.1 | |
XM_011513847.2 | 1543 | Missense Mutation | AAC,AAG | N,K 441 | XP_011512149.1 | |
XM_011513848.1 | 1543 | Missense Mutation | AAC,AAG | N,K 392 | XP_011512150.1 | |
XM_017008277.1 | 1543 | Missense Mutation | AAC,AAG | N,K 537 | XP_016863766.1 | |
XM_017008278.1 | 1543 | Missense Mutation | AAC,AAG | N,K 311 | XP_016863767.1 |