Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603135 | ||||||||||||||||||||
Literature Links: |
CUL2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CUL2 - cullin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198777.1 | 3858 | UTR 3 | NP_001185706.1 | |||
NM_001198778.1 | 3858 | UTR 3 | NP_001185707.1 | |||
NM_001198779.1 | 3858 | UTR 3 | NP_001185708.1 | |||
NM_001324375.1 | 3858 | UTR 3 | NP_001311304.1 | |||
NM_001324376.1 | 3858 | UTR 3 | NP_001311305.1 | |||
NM_003591.3 | 3858 | UTR 3 | NP_003582.2 |