Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300368 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC9A7 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SLC9A7 - solute carrier family 9 member A7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001257291.1 | Intron | NP_001244220.1 | ||||
NM_032591.2 | Intron | NP_115980.1 | ||||
XM_005272681.4 | Intron | XP_005272738.1 | ||||
XM_005272682.4 | Intron | XP_005272739.1 | ||||
XM_006724564.3 | Intron | XP_006724627.1 | ||||
XM_011543990.2 | Intron | XP_011542292.1 | ||||
XM_017029905.1 | Intron | XP_016885394.1 | ||||
XM_017029906.1 | Intron | XP_016885395.1 |
Set Membership: |
HapMap |