Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||
Literature Links: |
KIAA0226L PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
KIAA0226L - KIAA0226 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286761.1 | 2585 | UTR 3 | NP_001273690.1 | |||
NM_001286762.1 | 2585 | UTR 3 | NP_001273691.1 | |||
NM_001286763.1 | 2585 | UTR 3 | NP_001273692.1 | |||
NM_001286764.1 | 2585 | UTR 3 | NP_001273693.1 | |||
NM_001286765.1 | 2585 | UTR 3 | NP_001273694.1 | |||
NM_001286766.1 | 2585 | UTR 3 | NP_001273695.1 | |||
NM_025113.3 | 2585 | UTR 3 | NP_079389.2 |