Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGAGGCTTATCATCACCATGGAGA[C/T]ATCCTTCTTCAGCTTCTTCATCTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607884 MIM: 607885 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CKLF-CMTM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CKLF-CMTM1 - CKLF-CMTM1 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001202509.1 | 525 | Intron | NP_001189438.1 | |||
NM_001204098.1 | 525 | Intron | NP_001191027.1 | |||
NM_001204099.1 | 525 | Intron | NP_001191028.1 |
CMTM1 - CKLF like MARVEL transmembrane domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_052999.3 | 525 | Intron | NP_443725.3 | |||
NM_181268.2 | 525 | Intron | NP_851785.2 | |||
NM_181269.2 | 525 | Intron | NP_851786.1 | |||
NM_181270.2 | 525 | Intron | NP_851787.1 | |||
NM_181271.2 | 525 | Intron | NP_851788.1 | |||
NM_181272.2 | 525 | Intron | NP_851789.1 | |||
NM_181283.2 | 525 | Intron | NP_851800.1 | |||
NM_181296.2 | 525 | Intron | NP_851813.1 |
CMTM2 - CKLF like MARVEL transmembrane domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199317.1 | 525 | Intron | NP_001186246.1 | |||
NM_144673.2 | 525 | Missense Mutation | ACA,ATA | T,I 122 | NP_653274.1 |
Set Membership: |
HapMap Validated |