Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609768 MIM: 604532 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BLOC1S2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
BLOC1S2 - biogenesis of lysosomal organelles complex 1 subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001342.2 | Intron | NP_001001342.1 | ||||
NM_001282436.1 | Intron | NP_001269365.1 | ||||
NM_001282437.1 | Intron | NP_001269366.1 | ||||
NM_001282438.1 | Intron | NP_001269367.1 | ||||
NM_001282439.1 | Intron | NP_001269368.1 | ||||
NM_173809.4 | Intron | NP_776170.2 | ||||
XM_017016102.1 | Intron | XP_016871591.1 |
PKD2L1 - polycystin 2 like 1, transient receptor potential cation channel | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |