Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 104615 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MTUS2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
MTUS2 - microtubule associated tumor suppressor candidate 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC7A1 - solute carrier family 7 member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003045.4 | 6609 | UTR 3 | NP_003036.1 | |||
XM_005266507.3 | 6609 | Intron | XP_005266564.1 | |||
XM_017020713.1 | 6609 | UTR 3 | XP_016876202.1 | |||
XM_017020714.1 | 6609 | UTR 3 | XP_016876203.1 | |||
XM_017020715.1 | 6609 | UTR 3 | XP_016876204.1 | |||
XM_017020716.1 | 6609 | Intron | XP_016876205.1 |
Set Membership: |
HapMap |