Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAATCCATTTGTCTGGTAGTGTAG[C/G]ATAGAGTCCAGAAATAGATCCACAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613297 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101929977 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC101929977 - uncharacterized LOC101929977 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MARCH6 - membrane associated ring-CH-type finger 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270660.1 | Intron | NP_001257589.1 | ||||
NM_001270661.1 | Intron | NP_001257590.1 | ||||
NM_005885.3 | Intron | NP_005876.2 | ||||
XM_011513932.2 | Intron | XP_011512234.1 | ||||
XM_011513934.1 | Intron | XP_011512236.1 | ||||
XM_011513936.2 | Intron | XP_011512238.1 | ||||
XM_011513937.2 | Intron | XP_011512239.1 | ||||
XM_017008944.1 | Intron | XP_016864433.1 |
Set Membership: |
HapMap |