Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606959 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MPP6 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MPP6 - membrane palmitoylated protein 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303037.1 | Intron | NP_001289966.1 | ||||
NM_016447.3 | Intron | NP_057531.2 | ||||
XM_005249775.4 | Intron | XP_005249832.1 | ||||
XM_006715738.3 | Intron | XP_006715801.1 | ||||
XM_006715739.3 | Intron | XP_006715802.1 | ||||
XM_006715740.3 | Intron | XP_006715803.1 | ||||
XM_011515425.2 | Intron | XP_011513727.1 | ||||
XM_017012315.1 | Intron | XP_016867804.1 | ||||
XM_017012316.1 | Intron | XP_016867805.1 | ||||
XM_017012317.1 | Intron | XP_016867806.1 |
Set Membership: |
HapMap |