Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 173910 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PKD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PKD2 - polycystin 2, transient receptor potential cation channel | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000297.3 | 500 | Silent Mutation | GGA,GGG | G,G 140 | NP_000288.1 | |
XM_011532028.2 | 500 | Silent Mutation | GGA,GGG | G,G 140 | XP_011530330.1 | |
XM_011532029.1 | 500 | Intron | XP_011530331.1 | |||
XM_011532030.2 | 500 | Intron | XP_011530332.1 |