Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
17 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603272 MIM: 616698 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CNKSR1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CNKSR1 - connector enhancer of kinase suppressor of Ras 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FAM110D - family with sequence similarity 110 member D | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF593 - zinc finger protein 593 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015871.4 | 104 | Missense Mutation | CAG,CGG | Q,R 6 | NP_056955.2 | |
XM_017001398.1 | 104 | Missense Mutation | CAG,CGG | Q,R 6 | XP_016856887.1 |
Set Membership: |
HapMap |