Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609596 MIM: 601983 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EIF3K PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
EIF3K - eukaryotic translation initiation factor 3 subunit K | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300992.1 | Intron | NP_001287921.1 | ||||
NM_001308393.1 | Intron | NP_001295322.1 | ||||
NM_013234.3 | Intron | NP_037366.1 |
MAP4K1 - mitogen-activated protein kinase kinase kinase kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042600.2 | Intron | NP_001036065.1 | ||||
NM_007181.5 | Intron | NP_009112.1 | ||||
XM_011526403.1 | Intron | XP_011524705.1 | ||||
XM_011526404.1 | Intron | XP_011524706.1 | ||||
XM_017026231.1 | Intron | XP_016881720.1 |
Set Membership: |
HapMap |