Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609600 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF396 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZNF396 - zinc finger protein 396 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001322286.1 | 1461 | UTR 3 | NP_001309215.1 | |||
NM_001322290.1 | 1461 | UTR 3 | NP_001309219.1 | |||
NM_001322291.1 | 1461 | Intron | NP_001309220.1 | |||
NM_145756.2 | 1461 | UTR 3 | NP_665699.1 | |||
XM_006722432.3 | 1461 | UTR 3 | XP_006722495.1 | |||
XM_017025687.1 | 1461 | UTR 3 | XP_016881176.1 | |||
XM_017025688.1 | 1461 | UTR 3 | XP_016881177.1 | |||
XM_017025689.1 | 1461 | UTR 3 | XP_016881178.1 |
Set Membership: |
HapMap |