Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610388 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DEFB124 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
DEFB124 - defensin beta 124 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LINC00028 - long intergenic non-protein coding RNA 28 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
REM1 - RRAD and GEM like GTPase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014012.5 | 473 | Missense Mutation | CCT,GCT | P,A 59 | NP_054731.2 | |
XM_005260404.1 | 473 | Missense Mutation | CCT,GCT | P,A 59 | XP_005260461.1 | |
XM_011528795.1 | 473 | Missense Mutation | CCT,GCT | P,A 59 | XP_011527097.1 | |
XM_017027833.1 | 473 | Missense Mutation | CCT,GCT | P,A 59 | XP_016883322.1 |