Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
46 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602636 MIM: 606892 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPP1R8 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
PPP1R8 - protein phosphatase 1 regulatory subunit 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002713.3 | 547 | UTR 5 | NP_002704.1 | |||
NM_014110.4 | 547 | Intron | NP_054829.2 | |||
NM_138558.2 | 547 | Intron | NP_612568.1 | |||
XM_017001594.1 | 547 | Intron | XP_016857083.1 |
SCARNA1 - small Cajal body-specific RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STX12 - syntaxin 12 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap JSNP Validated |