Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 126453 MIM: 606142 | ||||||||||||||||||||
Literature Links: |
DRD5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DRD5 - dopamine receptor D5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000798.4 | 648 | Silent Mutation | GCC,GCT | A,A 84 | NP_000789.1 |
SLC2A9 - solute carrier family 2 member 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001290.1 | 648 | Intron | NP_001001290.1 | |||
NM_020041.2 | 648 | Intron | NP_064425.2 | |||
XM_006713968.3 | 648 | Intron | XP_006714031.1 | |||
XM_006713969.2 | 648 | Intron | XP_006714032.1 | |||
XM_011513856.2 | 648 | Intron | XP_011512158.1 | |||
XM_011513857.1 | 648 | Intron | XP_011512159.1 | |||
XM_011513858.1 | 648 | Intron | XP_011512160.1 | |||
XM_011513859.2 | 648 | Intron | XP_011512161.1 | |||
XM_011513860.2 | 648 | Intron | XP_011512162.1 | |||
XM_011513861.2 | 648 | Intron | XP_011512163.1 | |||
XM_011513862.2 | 648 | Intron | XP_011512164.1 | |||
XM_011513863.2 | 648 | Intron | XP_011512165.1 | |||
XM_011513864.2 | 648 | Intron | XP_011512166.1 | |||
XM_011513865.2 | 648 | Intron | XP_011512167.1 | |||
XM_011513866.2 | 648 | Intron | XP_011512168.1 | |||
XM_011513867.2 | 648 | Intron | XP_011512169.1 | |||
XM_011513868.2 | 648 | Intron | XP_011512170.1 | |||
XM_017008457.1 | 648 | Intron | XP_016863946.1 | |||
XM_017008458.1 | 648 | Intron | XP_016863947.1 | |||
XM_017008459.1 | 648 | Intron | XP_016863948.1 | |||
XM_017008460.1 | 648 | Intron | XP_016863949.1 |