Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGGAGTCGTACACACTGGCTCCAG[A/G]AGGAGGATACAAGCAATTCTGGGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 194555 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100379224 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC100379224 - uncharacterized LOC100379224 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF224 - zinc finger protein 224 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF225 - zinc finger protein 225 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321685.1 | Intron | NP_001308614.1 | ||||
NM_013362.3 | Intron | NP_037494.2 | ||||
XM_005259223.3 | Intron | XP_005259280.1 | ||||
XM_011527285.2 | Intron | XP_011525587.1 | ||||
XM_011527286.2 | Intron | XP_011525588.1 |
Set Membership: |
HapMap |