Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||
Phenotype: |
MIM: 606231 MIM: 615748 | ||||||||||||||||||||||||||
Literature Links: |
APPL2 PubMed Links | ||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese)
|
|||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
APPL2 - adaptor protein, phosphotyrosine interacting with PH domain and leucine zipper 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001251904.1 | 4168 | UTR 3 | NP_001238833.1 | |||
NM_001251905.1 | 4168 | UTR 3 | NP_001238834.1 | |||
NM_018171.3 | 4168 | UTR 3 | NP_060641.2 | |||
XM_006719472.1 | 4168 | UTR 3 | XP_006719535.1 | |||
XM_011538530.2 | 4168 | UTR 3 | XP_011536832.1 | |||
XM_011538531.2 | 4168 | UTR 3 | XP_011536833.1 | |||
XM_011538532.2 | 4168 | UTR 3 | XP_011536834.1 | |||
XM_017019551.1 | 4168 | UTR 3 | XP_016875040.1 | |||
XM_017019552.1 | 4168 | UTR 3 | XP_016875041.1 | |||
XM_017019553.1 | 4168 | UTR 3 | XP_016875042.1 | |||
XM_017019554.1 | 4168 | UTR 3 | XP_016875043.1 |
KIAA1033 - KIAA1033 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |