Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609169 MIM: 609969 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GAPDHS PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GAPDHS - glyceraldehyde-3-phosphate dehydrogenase, spermatogenic | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SBSN - suprabasin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166034.1 | Intron | NP_001159506.1 | ||||
NM_001166035.1 | Intron | NP_001159507.1 | ||||
NM_198538.3 | Intron | NP_940940.1 | ||||
XM_011526929.2 | Intron | XP_011525231.1 | ||||
XM_011526930.2 | Intron | XP_011525232.1 | ||||
XM_011526931.2 | Intron | XP_011525233.1 |