Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615417 MIM: 608356 MIM: 609146 MIM: 610176 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BET1L PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
BET1L - Bet1 golgi vesicular membrane trafficking protein like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098787.1 | 1733 | UTR 3 | NP_001092257.1 | |||
NM_016526.4 | 1733 | UTR 3 | NP_057610.2 |
MIR6743 - microRNA 6743 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ODF3 - outer dense fiber of sperm tails 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RIC8A - RIC8 guanine nucleotide exchange factor A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SCGB1C1 - secretoglobin family 1C member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |