Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602381 MIM: 616496 MIM: 601512 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NAB2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NAB2 - NGFI-A binding protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NEMP1 - nuclear envelope integral membrane protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STAT6 - signal transducer and activator of transcription 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001178078.1 | 2901 | Silent Mutation | NP_001171549.1 | |||
NM_001178079.1 | 2901 | Silent Mutation | NP_001171550.1 | |||
NM_001178080.1 | 2901 | Silent Mutation | NP_001171551.1 | |||
NM_001178081.1 | 2901 | Silent Mutation | NP_001171552.1 | |||
NM_003153.4 | 2901 | Silent Mutation | NP_003144.3 | |||
XM_006719575.2 | 2901 | Silent Mutation | XP_006719638.1 | |||
XM_011538703.2 | 2901 | Silent Mutation | XP_011537005.1 | |||
XM_011538704.2 | 2901 | Silent Mutation | XP_011537006.1 | |||
XM_011538705.2 | 2901 | Silent Mutation | XP_011537007.1 | |||
XM_011538706.2 | 2901 | Silent Mutation | XP_011537008.1 | |||
XM_011538707.2 | 2901 | Silent Mutation | XP_011537009.1 | |||
XM_011538708.2 | 2901 | Missense Mutation | XP_011537010.1 | |||
XM_011538709.2 | 2901 | Silent Mutation | XP_011537011.1 |