Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612085 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RASL12 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
RASL12 - RAS like family 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001307930.1 | Intron | NP_001294859.1 | ||||
NM_016563.3 | Intron | NP_057647.1 | ||||
XM_005254434.4 | Intron | XP_005254491.1 | ||||
XM_011521660.2 | Intron | XP_011519962.1 | ||||
XM_017022296.1 | Intron | XP_016877785.1 |
SLC51B - solute carrier family 51 beta subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178859.3 | Intron | NP_849190.2 | ||||
XM_005254159.4 | Intron | XP_005254216.1 |
Set Membership: |
HapMap |