Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTGTGTGTTGAAGTTGCTAAGATC[G/T]ACAGTGAGAACAAATCTTCTAATCT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607117 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100287015 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LOC100287015 - uncharacterized LOC100287015 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MCPH1 - microcephalin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172574.1 | Intron | NP_001166045.1 | ||||
NM_001172575.1 | Intron | NP_001166046.1 | ||||
NM_001322042.1 | Intron | NP_001308971.1 | ||||
NM_001322043.1 | Intron | NP_001308972.1 | ||||
NM_001322045.1 | Intron | NP_001308974.1 | ||||
NM_024596.4 | Intron | NP_078872.2 | ||||
XM_011534755.2 | Intron | XP_011533057.1 | ||||
XM_011534756.2 | Intron | XP_011533058.1 | ||||
XM_011534757.2 | Intron | XP_011533059.1 | ||||
XM_011534758.2 | Intron | XP_011533060.1 | ||||
XM_011534759.2 | Intron | XP_011533061.1 | ||||
XM_011534760.2 | Intron | XP_011533062.1 | ||||
XM_017013829.1 | Intron | XP_016869318.1 | ||||
XM_017013830.1 | Intron | XP_016869319.1 | ||||
XM_017013831.1 | Intron | XP_016869320.1 | ||||
XM_017013832.1 | Intron | XP_016869321.1 | ||||
XM_017013833.1 | Intron | XP_016869322.1 | ||||
XM_017013834.1 | Intron | XP_016869323.1 |
Set Membership: |
HapMap |