Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603193 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATP5G2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ATP5G2 - ATP synthase, H+ transporting, mitochondrial Fo complex subunit C2 (subunit 9) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002031.2 | Intron | NP_001002031.1 | ||||
NM_005176.5 | Intron | NP_005167.2 | ||||
XM_017019460.1 | Intron | XP_016874949.1 | ||||
XM_017019461.1 | Intron | XP_016874950.1 | ||||
XM_017019462.1 | Intron | XP_016874951.1 |
Set Membership: |
HapMap |