Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||
Phenotype: |
MIM: 603127 MIM: 179618 | ||||||||||||||||||||||||||
Literature Links: |
GAS7 PubMed Links | ||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
GAS7 - growth arrest specific 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130831.1 | 6688 | UTR 3 | NP_001124303.1 | |||
NM_003644.2 | 6688 | UTR 3 | NP_003635.2 | |||
NM_201432.1 | 6688 | UTR 3 | NP_958836.1 | |||
NM_201433.1 | 6688 | UTR 3 | NP_958839.1 | |||
XM_005256831.3 | 6688 | UTR 3 | XP_005256888.1 | |||
XM_005256832.3 | 6688 | UTR 3 | XP_005256889.1 | |||
XM_005256833.3 | 6688 | UTR 3 | XP_005256890.1 | |||
XM_005256834.3 | 6688 | UTR 3 | XP_005256891.1 | |||
XM_005256835.3 | 6688 | UTR 3 | XP_005256892.1 | |||
XM_011524044.2 | 6688 | UTR 3 | XP_011522346.1 | |||
XM_017025239.1 | 6688 | UTR 3 | XP_016880728.1 | |||
XM_017025240.1 | 6688 | UTR 3 | XP_016880729.1 | |||
XM_017025241.1 | 6688 | UTR 3 | XP_016880730.1 | |||
XM_017025242.1 | 6688 | UTR 3 | XP_016880731.1 |
RCVRN - recoverin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |