Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600632 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
OPCML PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
OPCML - opioid binding protein/cell adhesion molecule like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012393.2 | 18637 | Intron | NP_001012393.1 | |||
NM_001319103.1 | 18637 | Intron | NP_001306032.1 | |||
NM_001319104.1 | 18637 | Intron | NP_001306033.1 | |||
NM_001319105.1 | 18637 | Intron | NP_001306034.1 | |||
NM_001319106.1 | 18637 | Intron | NP_001306035.1 | |||
NM_002545.4 | 18637 | Intron | NP_002536.1 | |||
XM_006718846.2 | 18637 | UTR 3 | XP_006718909.1 | |||
XM_011542856.2 | 18637 | UTR 3 | XP_011541158.1 |
Set Membership: |
HapMap |