Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF841 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ZNF841 - zinc finger protein 841 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136499.1 | Intron | NP_001129971.1 | ||||
NM_001321349.1 | Intron | NP_001308278.1 | ||||
XM_011526833.2 | Intron | XP_011525135.1 | ||||
XM_017026652.1 | Intron | XP_016882141.1 | ||||
XM_017026653.1 | Intron | XP_016882142.1 | ||||
XM_017026654.1 | Intron | XP_016882143.1 | ||||
XM_017026655.1 | Intron | XP_016882144.1 | ||||
XM_017026656.1 | Intron | XP_016882145.1 | ||||
XM_017026657.1 | Intron | XP_016882146.1 | ||||
XM_017026658.1 | Intron | XP_016882147.1 | ||||
XM_017026659.1 | Intron | XP_016882148.1 | ||||
XM_017026660.1 | Intron | XP_016882149.1 | ||||
XM_017026661.1 | Intron | XP_016882150.1 | ||||
XM_017026662.1 | Intron | XP_016882151.1 | ||||
XM_017026663.1 | Intron | XP_016882152.1 |
Set Membership: |
HapMap |