Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 191050 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
WARS PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
WARS - tryptophanyl-tRNA synthetase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WDR25 - WD repeat domain 25 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161476.1 | 276 | Missense Mutation | AAA,AGA | K,R 59 | NP_001154948.1 | |
NM_024515.4 | 276 | Missense Mutation | AAA,AGA | K,R 59 | NP_078791.3 | |
XM_005268056.4 | 276 | Missense Mutation | AAA,AGA | K,R 59 | XP_005268113.1 | |
XM_005268057.3 | 276 | Intron | XP_005268114.1 | |||
XM_005268058.4 | 276 | Intron | XP_005268115.1 | |||
XM_011537150.2 | 276 | Missense Mutation | AAA,AGA | K,R 59 | XP_011535452.1 | |
XM_011537151.2 | 276 | Intron | XP_011535453.2 | |||
XM_017021639.1 | 276 | Intron | XP_016877128.1 |
Set Membership: |
HapMap |